ENA387772605 Enamine

2-(1,4-diazepan-1-yl)-1H-1,3-benzodiazole

  • MDL number MFCD09701682
  • Cas Number : 692724-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119593

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119593
Availability details for
Quantity
  • Availability
  • Add to Cart