ENA387772608 Enamine

4-(chloromethyl)-2-(oxolan-2-yl)-1,3-thiazole

  • MDL number MFCD21193532
  • Cas Number : 1423028-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119597
Availability details for
Quantity
  • Availability
  • Add to Cart