ENA387772609 Enamine

3-methyl-4-(thiophene-2-amido)benzoic acid

  • MDL number MFCD06623944
  • Cas Number : 827002-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119598
Availability details for
Quantity
  • Availability
  • Add to Cart