ENA387772612 Enamine

4-[(4-bromo-1H-pyrazol-1-yl)methyl]-5-chloro-1,3-dimethyl-1H-pyrazole

  • MDL number MFCD12823050
  • Cas Number : 1178837-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52...54
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119603

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119603
Availability details for
Quantity
  • Availability
  • Add to Cart