ENA387772613 Enamine

2-(2-fluorophenyl)-5-methyl-1,3,4-oxadiazole

  • MDL number MFCD01617922
  • Cas Number : 345994-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119604
Availability details for
Quantity
  • Availability
  • Add to Cart