ENA387772614 Enamine

{4-[(dimethylamino)methyl]-3-fluorophenyl}methanamine

  • MDL number MFCD09803129
  • Cas Number : 1016711-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116628
Availability details for
Quantity
  • Availability
  • Add to Cart