ENA387772622 Enamine

4-(hydrazinesulfonyl)benzoic acid

  • MDL number MFCD00025109
  • Cas Number : 6391-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119619
Availability details for
Quantity
  • Availability
  • Add to Cart