ENA387772623 Enamine

2-(bromomethyl)-5-nitrobenzoic acid

  • MDL number MFCD22053120
  • Cas Number : 21626-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119622
Availability details for
Quantity
  • Availability
  • Add to Cart