ENA387772626 Enamine

(4-aminopyridin-2-yl)methanol

  • MDL number MFCD11100581
  • Cas Number : 100114-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116645
Availability details for
Quantity
  • Availability
  • Add to Cart