ENA387772630 Enamine

1-ethyl-3-oxo-1,2,3,4-tetrahydroquinoxaline-6-carboxylic acid

  • MDL number MFCD16113613
  • Cas Number : 1031655-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119634
Availability details for
Quantity
  • Availability
  • Add to Cart