ENA387772635 Enamine

1-(2-aminophenyl)-2-methylpropan-1-one

  • MDL number MFCD20426719
  • Cas Number : 27309-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119641

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119641
Availability details for
Quantity
  • Availability
  • Add to Cart