ENA387772636 Enamine

3-[(2,4-difluorophenyl)methyl]-8-azabicyclo[3.2.1]octan-3-ol

  • MDL number MFCD18592189
  • Cas Number : 1405208-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119642
Availability details for
Quantity
  • Availability
  • Add to Cart