ENA387772649 Enamine

2-(1,2,3,4-tetrahydroisoquinolin-2-yl)quinolin-6-amine

  • MDL number MFCD13698780
  • Cas Number : 1154583-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-119692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119692
Availability details for
Quantity
  • Availability
  • Add to Cart