ENA387772661 Enamine

1-(1-ethyl-1H-pyrazol-4-yl)-2,2,2-trifluoroethan-1-one

  • MDL number MFCD10699048
  • Cas Number : 1174848-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117599

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117599
Availability details for
Quantity
  • Availability
  • Add to Cart