ENA387772666 Enamine

ethyl 3-bromo-5-(chlorosulfonyl)benzoate

  • MDL number MFCD12169538
  • Cas Number : 1155912-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117628

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117628
Availability details for
Quantity
  • Availability
  • Add to Cart