ENA387772669 Enamine

1-(2,6-dichlorophenyl)propan-2-amine hydrochloride

  • MDL number MFCD22628447
  • Cas Number : 1423033-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119742

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119742
Availability details for
Quantity
  • Availability
  • Add to Cart