ENA387772672 Enamine

6-(tert-butoxy)pyridine-2-carboximidamide

  • MDL number MFCD20367828
  • Cas Number : 1339092-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119779
Availability details for
Quantity
  • Availability
  • Add to Cart