ENA387772674 Enamine

3-amino-1,2-benzoxazole-5-carbaldehyde

  • MDL number MFCD18804968
  • Cas Number : 648449-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119783
Availability details for
Quantity
  • Availability
  • Add to Cart