ENA387772680 Enamine

5-bromo-2,3-dihydro-1-benzofuran-3-ol

  • MDL number MFCD21295500
  • Cas Number : 5590-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119789
Availability details for
Quantity
  • Availability
  • Add to Cart