ENA387772689 Enamine

3-(2,2-dimethylmorpholin-4-yl)-N'-hydroxy-3-oxopropanimidamide

  • MDL number MFCD12081007
  • Cas Number : 1158093-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119799

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119799
Availability details for
Quantity
  • Availability
  • Add to Cart