ENA387772691 Enamine

4-[(propan-2-yl)carbamoyl]benzoic acid

  • MDL number MFCD01682706
  • Cas Number : 779-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119808

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119808
Availability details for
Quantity
  • Availability
  • Add to Cart