ENA387772693 Enamine

2-(5-methylpyrimidin-2-yl)ethan-1-amine

  • MDL number MFCD10697250
  • Cas Number : 933682-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119810
Availability details for
Quantity
  • Availability
  • Add to Cart