ENA387772697 Enamine

3-chloro-4-methyl-4H-1,2,4-triazole

  • MDL number MFCD09995739
  • Cas Number : 56616-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119818

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-119818
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart