ENA387772698 Enamine

1-[(1-phenyl-1H-pyrazol-4-yl)methyl]-1H-pyrazol-5-amine

  • MDL number MFCD09737207
  • Cas Number : 1052554-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119822
Availability details for
Quantity
  • Availability
  • Add to Cart