ENA387772700 Enamine

5-(difluoromethyl)-1-(2-methylpropyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD17240902
  • Cas Number : 1339834-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119824
Availability details for
Quantity
  • Availability
  • Add to Cart