ENA387772701 Enamine

2-(2-methoxybenzoyl)pyridine

  • MDL number MFCD07699223
  • Cas Number : 22945-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119826
Availability details for
Quantity
  • Availability
  • Add to Cart