ENA387772707 Enamine

5-(difluoromethyl)-1-(2-methylpropyl)-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD22628453
  • Cas Number : 1423023-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119832
Availability details for
Quantity
  • Availability
  • Add to Cart