ENA387772716 Enamine

1-benzyl-3-methylpiperidin-3-ol

  • MDL number MFCD22628457
  • Cas Number : 6560-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119851
Availability details for
Quantity
  • Availability
  • Add to Cart