ENA387772718 Enamine

methyl 5-amino-6-methylpyridine-2-carboxylate

  • MDL number MFCD20715759
  • Cas Number : 343786-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117806
Availability details for
Quantity
  • Availability
  • Add to Cart