ENA387772724 Enamine

ethyl 2-ethyl-4-methyl-3-oxopentanoate

  • MDL number MFCD00511277
  • Cas Number : 54285-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119895

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119895
Availability details for
Quantity
  • Availability
  • Add to Cart