ENA387772728 Enamine

5-methanesulfonyl-2,3-dimethylbenzoic acid

  • MDL number MFCD22628462
  • Cas Number : 1423028-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119901
Availability details for
Quantity
  • Availability
  • Add to Cart