ENA387772733 Enamine

(4-cyclopropyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD22628360
  • Cas Number : 135207-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117933
Availability details for
Quantity
  • Availability
  • Add to Cart