ENA387772735 Enamine

1-(1,3-thiazol-2-yl)-1H-pyrazol-3-amine

  • MDL number MFCD14594762
  • Cas Number : 1251131-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-119910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119910
Availability details for
Quantity
  • Availability
  • Add to Cart