ENA387772737 Enamine

4-(aminomethyl)-1-benzylpyrrolidin-2-one hydrochloride

  • MDL number MFCD09743242
  • Cas Number : 1177279-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119912
Availability details for
Quantity
  • Availability
  • Add to Cart