ENA387772738 Enamine

1,2,4-oxadiazole-3-carboxamide

  • MDL number MFCD19208306
  • Cas Number : 39512-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119913
Availability details for
Quantity
  • Availability
  • Add to Cart