ENA387772748 Enamine

3-chloro-4-hydroxy-N-(2-methoxyethyl)benzamide

  • MDL number MFCD11130668
  • Cas Number : 1019466-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118125
Availability details for
Quantity
  • Availability
  • Add to Cart