ENA387772756 Enamine

2-[3-(methylcarbamoyl)phenoxy]acetic acid

  • MDL number MFCD10037128
  • Cas Number : 1018600-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108487
Availability details for
Quantity
  • Availability
  • Add to Cart