ENA387772768 Enamine

2-(3-phenyl-1,2,4-oxadiazol-5-yl)propan-1-amine

  • MDL number MFCD16839892
  • Cas Number : 1423028-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119969

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119969
Availability details for
Quantity
  • Availability
  • Add to Cart