ENA387772772 Enamine

methyl 3-amino-5-iodobenzoate

  • MDL number MFCD08447271
  • Cas Number : 217314-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118420

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-118420
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart