ENA387772773 Enamine

4-aminothiane-4-carboxylic acid

  • MDL number MFCD06656113
  • Cas Number : 39124-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108712

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108712
Availability details for
Quantity
  • Availability
  • Add to Cart