ENA387772777 Enamine

2-ethyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD00705111
  • Cas Number : 56844-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119979

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119979
Availability details for
Quantity
  • Availability
  • Add to Cart