ENA387772779 Enamine

1-bromo-3-[(methylsulfanyl)methyl]benzene

  • MDL number MFCD14644218
  • Cas Number : 76354-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118551

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-118551
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart