ENA387772781 Enamine

3-[4-(4-chlorophenyl)piperazin-1-yl]propanenitrile

  • MDL number MFCD11164462
  • Cas Number : 79322-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120007

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120007
Availability details for
Quantity
  • Availability
  • Add to Cart