ENA387772783 Enamine

1-N,1-N-dimethyl-4-N-(1H-pyrrol-2-ylmethyl)benzene-1,4-diamine

  • MDL number MFCD16734468
  • Cas Number : 1250358-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118634

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118634
Availability details for
Quantity
  • Availability
  • Add to Cart