ENA387772787 Enamine

(1S)-1-(6-methylpyridin-2-yl)ethan-1-ol

  • MDL number MFCD18828034
  • Cas Number : 204244-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-44
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120037
Availability details for
Quantity
  • Availability
  • Add to Cart