ENA387772792 Enamine

4-ethynyl-2,3,5,6-tetrafluoroaniline

  • MDL number MFCD20709982
  • Cas Number : 61794-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120045
Availability details for
Quantity
  • Availability
  • Add to Cart