ENA387772798 Enamine

2-(difluoromethyl)-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD11193511
  • Cas Number : 891831-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120056
Availability details for
Quantity
  • Availability
  • Add to Cart