ENA387772799 Enamine

2-methyl-4-oxocyclohexane-1-carboxylic acid

  • MDL number MFCD20641494
  • Cas Number : 115989-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120057

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-120057
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart