ENA387772802 Enamine

3,5-dimethyl-4-(pyrrolidin-2-yl)-1H-pyrazole

  • MDL number MFCD22068366
  • Cas Number : 1367696-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120060

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120060
Availability details for
Quantity
  • Availability
  • Add to Cart