ENA387772806 Enamine

4-(5-chloro-3-methyl-1H-pyrazol-1-yl)benzonitrile

  • MDL number MFCD21667611
  • Cas Number : 1423029-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120064
Availability details for
Quantity
  • Availability
  • Add to Cart